missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-boronic acid, 98%, Thermo Scientific Chemicals

23.00 € - 222.00 €

Chemical Identifiers

CAS 206551-43-1
Molecular Formula C6H7BO3S
Molecular Weight (g/mol) 169.989
MDL Number MFCD01075681
InChI Key DCNMATSPQKWETQ-UHFFFAOYSA-N
Synonym 5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
PubChem CID 4143377
IUPAC Name (5-acetylthiophen-2-yl)boronic acid
SMILES B(C1=CC=C(S1)C(=O)C)(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328216
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L15221.03
1 g
23.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11338216
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L15221.06
5 g
68.40 €
5g
Please to purchase this item. Need a web account? Register with us today!
11348216
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L15221.14
25 g
222.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

206551-43-1
169.989
DCNMATSPQKWETQ-UHFFFAOYSA-N
4143377
B(C1=CC=C(S1)C(=O)C)(O)O
C6H7BO3S
MFCD01075681
5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
(5-acetylthiophen-2-yl)boronic acid

Specifications

206551-43-1
C6H7BO3S
8681302
DCNMATSPQKWETQ-UHFFFAOYSA-N
(5-acetylthiophen-2-yl)boronic acid
4143377
98%
142°C to 148°C
MFCD01075681
5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
B(C1=CC=C(S1)C(=O)C)(O)O
169.989
170
5-Acetylthiophene-2-boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only