missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chloro-3-nitrophenyl)-2-furoyl chloride, Thermo Scientific™

Product Code. 11305409
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638160-09-5
Molecular Formula C11H5Cl2NO4
Molecular Weight (g/mol) 286.064
MDL Number MFCD02258014
InChI Key YASHGRDOYUBRTA-UHFFFAOYSA-N
Synonym 5-4-chloro-3-nitrophenyl furan-2-carbonyl chloride, 5-4-chloro-3-nitrophenyl-2-furoyl chloride, acmc-20am8v, 2-furancarbonylchloride, 5-4-chloro-3-nitrophenyl, 5-4-chloro-3-nitro-phenyl-furan-2-carbonyl chloride
PubChem CID 2772077
IUPAC Name 5-(4-chloro-3-nitrophenyl)furan-2-carbonyl chloride
SMILES C1=CC(=C(C=C1C2=CC=C(O2)C(=O)Cl)[N+](=O)[O-])Cl

Specifications

Quantity 5 g
UN Number UN3261
Formula Weight 286.07
Chemical Name or Material 5-(4-Chloro-3-nitrophenyl)-2-furoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.