missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Chloro-4-nitrophenyl)-2-furoyl chloride, Thermo Scientific™

Product Code. 11365399
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380594-11-6
Molecular Formula C11H5Cl2NO4
Molecular Weight (g/mol) 286.064
MDL Number MFCD02258011
InChI Key BQTYBZIOLAXUSL-UHFFFAOYSA-N
Synonym 5-2-chloro-4-nitrophenyl furan-2-carbonyl chloride, 5-2-chloro-4-nitrophenyl-2-furoyl chloride, 5-2-chloro-4-nitro-phenyl-furan-2-carbonyl chloride, acmc-1ctg9, 5-2-chloro-4-nitrophenyl-2-furoylchloride
PubChem CID 4484926
IUPAC Name 5-(2-chloro-4-nitrophenyl)furan-2-carbonyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)C2=CC=C(O2)C(=O)Cl

Specifications

Quantity 5 g
UN Number UN3261
Formula Weight 286.07
Chemical Name or Material 5-(2-Chloro-4-nitrophenyl)-2-furoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'