missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ Rotary Valves with Omnifit Cap System

Rotary Valves with Omnifit Cap System features a PTFE body and PCTFE rotor that offers excellent chemical resistance.

Brand:  Diba Omnifit™ 21939-20

 View more versions of this product

Product Code. 15893035

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Pressure rated up to 50 psi (3.4 bar).
  • Rotary valves have a 1.5 mm bore and accept tubing sizes between 1 and 4 mm OD.
  • Select from various configurations to suit your application.
Specifications

Specifications

4 Way Valve 90°
1.5 mm
1 to 4 mm
Omnifit Cap System or 1/4 in.-28 UNF(F)
4 Ports
0.06 in.
0.04 in. to 0.16 in.
50 psi, 3.4 bar
4-way Valve
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers