missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ Rotary Valves with Omnifit Cap System

Rotary Valves with Omnifit Cap System features a PTFE body and PCTFE rotor that offers excellent chemical resistance.

Brand:  Diba Omnifit™ 21939-22

 View more versions of this product

Product Code. 15872585

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Pressure rated up to 50 psi (3.4 bar).
  • Rotary valves have a 1.5 mm bore and accept tubing sizes between 1 and 4 mm OD.
  • Select from various configurations to suit your application.
Specifications

Specifications

4 Way Valve 2 x 90°
1.5 mm
1 to 4 mm
Omnifit Cap System or 1/4 in.-28 UNF(F)
4 Ports
0.06 in.
0.04 in. to 0.16 in.
50 psi, 3.4 bar
4-way Valve
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.