missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetophenone, 98+%, Thermo Scientific Chemicals

Product Code. 11412957
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 709-63-7
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00000401
InChI Key HHAISVSEJFEWBZ-UHFFFAOYSA-N
Synonym 4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
PubChem CID 69731
IUPAC Name 1-[4-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 29°C to 35°C
Density 0.924
Boiling Point 83°C to 85°C (9 mmHg)
Flash Point 84°C (183°F)
Odor Characteristic
Refractive Index 1.462
Quantity 100 g
Beilstein 1870425
Formula Weight 188.15
Percent Purity ≥98%
Chemical Name or Material 4'-(Trifluoromethyl)acetophenone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.