Learn More
4-tert-Butylbenzyl bromide, 97%, Thermo Scientific Chemicals
30.00 € - 96.90 €
Chemical Identifiers
CAS | 18880-00-7 |
---|---|
Molecular Formula | C11H15Br |
Molecular Weight (g/mol) | 227.145 |
MDL Number | MFCD00000180 |
InChI Key | QZNQSIHCDAGZIA-UHFFFAOYSA-N |
Synonym | 4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide |
PubChem CID | 87836 |
IUPAC Name | 1-(bromomethyl)-4-tert-butylbenzene |
SMILES | CC(C)(C)C1=CC=C(C=C1)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421630
|
Thermo Scientific Alfa Aesar
H26991.06 |
5 g |
30.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431630
|
Thermo Scientific Alfa Aesar
H26991.14 |
25 g |
96.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18880-00-7 | |
227.145 | |
QZNQSIHCDAGZIA-UHFFFAOYSA-N | |
87836 | |
CC(C)(C)C1=CC=C(C=C1)CBr |
C11H15Br | |
MFCD00000180 | |
4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide | |
1-(bromomethyl)-4-tert-butylbenzene |
Specifications
18880-00-7 | |
1.236 | |
>110°C (230°F) | |
1.5465 | |
UN3265 | |
4-tert-butylbenzyl bromide, p-tert-butylbenzyl bromide, 1-bromomethyl-4-tert-butyl benzene, 4-tert-butylbenzylbromide, 1-bromomethyl-4-tert-butylbenzene, 4-tert-butyl benzylbromide, p-t-butylbenzyl bromide, 1-bromomethyl-4-1,1-dimethylethyl benzene, 4-t-butylbenzylbromide, 4-tert-buthylbenzyl bromide | |
CC(C)(C)C1=CC=C(C=C1)CBr | |
227.145 | |
227.14 | |
4-tert-Butylbenzyl bromide |
8°C to 12°C | |
93°C to 94°C (1.5 mmHg) | |
C11H15Br | |
MFCD00000180 | |
471674 | |
QZNQSIHCDAGZIA-UHFFFAOYSA-N | |
1-(bromomethyl)-4-tert-butylbenzene | |
87836 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 242-643-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only