Learn More
4-(tert-Butoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
43.30 € - 133.00 €
Chemical Identifiers
CAS | 850568-54-6 |
---|---|
Molecular Formula | C11H15BO4 |
Molecular Weight (g/mol) | 222.047 |
MDL Number | MFCD03411946 |
InChI Key | QMVMDYSTJSUDKC-UHFFFAOYSA-N |
Synonym | 4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid |
PubChem CID | 2773301 |
IUPAC Name | [4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid |
SMILES | B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O |
Description
Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Chemical Identifiers
850568-54-6 | |
222.047 | |
QMVMDYSTJSUDKC-UHFFFAOYSA-N | |
2773301 | |
B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O |
C11H15BO4 | |
MFCD03411946 | |
4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid | |
[4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid |
Specifications
850568-54-6 | |
C11H15BO4 | |
4-tert-butoxycarbonyl phenylboronic acid, 4-tert-butoxycarbonyl phenyl boronic acid, 4-t-butoxycarbonyl phenylboronic acid, tert-butyl 4-boronobenzoate, 4-t-butoxycarbonylphenylboronic acid, 4-boc-phenylboronic acid, 4-tert-butoxycarbonyl benzeneboronic acid, benzoic acid,4-borono-, 1,1-dimethylethyl ester, 4-t-butoxycarbonyl phenylboronicacid | |
QMVMDYSTJSUDKC-UHFFFAOYSA-N | |
[4-[(2-methylpropan-2-yl)oxycarbonyl]phenyl]boronic acid | |
2773301 | |
95% |
100°C to 106°C | |
MFCD03411946 | |
Slightly soluble in water. | |
B(C1=CC=C(C=C1)C(=O)OC(C)(C)C)(O)O | |
222.047 | |
222.05 | |
4-(tert-Butoxycarbonyl)benzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.