missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcarbamoylbenzeneboronic acid, 97%, Thermo Scientific™

66.00 € - 215.00 €

Chemical Identifiers

CAS 330793-45-8
Molecular Formula C13H12BNO3
Molecular Weight (g/mol) 241.053
MDL Number MFCD04115683
InChI Key DAUSGSRIYCUJDK-UHFFFAOYSA-N
Synonym 4-phenylcarbamoyl phenyl boronic acid, 4-phenylcarbamoyl phenylboronic acid, 4-phenylaminocarbonylphenylboronic acid, 4-phenylaminocarbonylphenyl boronic acid, 4-phenylaminocarbonyl benzeneboronic acid, 4-phenylamino-1-carbonyl phenyl boronic acid, n-phenyl 4-boronobenzamide, acmc-1afql, 4-phenylcarbamoylbenzeneboronic acid
PubChem CID 11096747
IUPAC Name [4-(phenylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15489917
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53159.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15479917
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53159.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

330793-45-8
241.053
DAUSGSRIYCUJDK-UHFFFAOYSA-N
11096747
B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2)(O)O
C13H12BNO3
MFCD04115683
4-phenylcarbamoyl phenyl boronic acid, 4-phenylcarbamoyl phenylboronic acid, 4-phenylaminocarbonylphenylboronic acid, 4-phenylaminocarbonylphenyl boronic acid, 4-phenylaminocarbonyl benzeneboronic acid, 4-phenylamino-1-carbonyl phenyl boronic acid, n-phenyl 4-boronobenzamide, acmc-1afql, 4-phenylcarbamoylbenzeneboronic acid
[4-(phenylcarbamoyl)phenyl]boronic acid

Specifications

330793-45-8
C13H12BNO3
4-phenylcarbamoyl phenyl boronic acid, 4-phenylcarbamoyl phenylboronic acid, 4-phenylaminocarbonylphenylboronic acid, 4-phenylaminocarbonylphenyl boronic acid, 4-phenylaminocarbonyl benzeneboronic acid, 4-phenylamino-1-carbonyl phenyl boronic acid, n-phenyl 4-boronobenzamide, acmc-1afql, 4-phenylcarbamoylbenzeneboronic acid
B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2)(O)O
241.053
241.05
4-Phenylcarbamoylbenzeneboronic acid
210°C to 216°C
MFCD04115683
DAUSGSRIYCUJDK-UHFFFAOYSA-N
[4-(phenylcarbamoyl)phenyl]boronic acid
11096747
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications