missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, 97%, Thermo Scientific Chemicals

Product Code. 10589803
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1689-82-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002330
InChI Key JTSBGMZPPPULTA-UHFFFAOYSA-N
Synonym C.I. 11800, 4-Hydroxyazobenzene, Solvent Yellow 7
PubChem CID 5354198
IUPAC Name 4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES O=C1C=CC(C=C1)=NNC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 16, 96
Infrared Spectrum Authentic
Formula Weight 198.22
Percent Purity ≥96.0%
Packaging Glass bottle
Color Brown
Quantity 100 g
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Phenylazophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Phenylazophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'