missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxypiperidine, 98%, Thermo Scientific Chemicals

Product Code. 11489750
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3202-33-3
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.25
MDL Number MFCD04114971
InChI Key KBYPITRKIJKGMD-UHFFFAOYSA-N
Synonym piperidine, 4-phenoxy, unii-aof4m4l61z, 4-phenoxy-piperidine, aof4m4l61z, piperidine, 4-phenoxy-, hydrochloride, 4-phenoxy piperidine, acmc-209hq3
PubChem CID 18878
IUPAC Name 4-phenoxypiperidine
SMILES C1CC(CCN1)OC1=CC=CC=C1

Specifications

Boiling Point 107°C to 108°C (0.1 mmHg)
Quantity 1 g
Formula Weight 177.25
Percent Purity 99%
Chemical Name or Material 4-Phenoxypiperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'