missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)phenylboronic acid, 97%, Thermo Scientific™

91.80 € - 148.00 €

Chemical Identifiers

CAS 397843-61-7
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.05
MDL Number MFCD09966155
InChI Key IXDHTLPOOZIZJX-UHFFFAOYSA-N
Synonym 4-phenoxymethyl phenylboronic acid, 4-phenoxymethyl phenyl boronic acid, acmc-1aegd, 4-phenoxymethyl phenyl boronicacid
PubChem CID 10955308
IUPAC Name [4-(phenoxymethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=C(COC2=CC=CC=C2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10183904
View Documents
Thermo Scientific Maybridge
CC63712CB
250 mg Amber Glass Bottle
91.80 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10557294
View Documents
Thermo Scientific Maybridge
CC63712DA
1 g Amber Glass Bottle
148.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

397843-61-7
228.05
IXDHTLPOOZIZJX-UHFFFAOYSA-N
10955308
OB(O)C1=CC=C(COC2=CC=CC=C2)C=C1
C13H13BO3
MFCD09966155
4-phenoxymethyl phenylboronic acid, 4-phenoxymethyl phenyl boronic acid, acmc-1aegd, 4-phenoxymethyl phenyl boronicacid
[4-(phenoxymethyl)phenyl]boronic acid

Specifications

397843-61-7
100.0
C13H13BO3
4-phenoxymethyl phenylboronic acid, 4-phenoxymethyl phenyl boronic acid, acmc-1aegd, 4-phenoxymethyl phenyl boronicacid
OB(O)C1=CC=C(COC2=CC=CC=C2)C=C1
228.05
228.05
4-(Phenoxymethyl)phenylboronic acid
96.5
97%
MFCD09966155
IXDHTLPOOZIZJX-UHFFFAOYSA-N
[4-(phenoxymethyl)phenyl]boronic acid
10955308
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.