Learn More
4-n-Propylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
44.00 € - 153.00 €
Chemical Identifiers
CAS | 146949-07-7 |
---|---|
Molecular Formula | C9H11ClO2S |
Molecular Weight (g/mol) | 218.70 |
MDL Number | MFCD00041508 |
InChI Key | LEFGAGRZHLNPLS-UHFFFAOYSA-N |
Synonym | 4-n-propylbenzenesulfonyl chloride, 4-propylbenzene-1-sulfonyl chloride, 4-propyl-benzenesulfonylchloride, 4-propyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-propyl, p-propylbenzenesulfonyl chloride, 4-n-propylbenzenesulfonylchloride, acmc-209cxt, 4-propylphenylsulfonyl chloride, 4-propylphenylsulphonyl chloride |
PubChem CID | 2737222 |
IUPAC Name | 4-propylbenzenesulfonyl chloride |
SMILES | CCCC1=CC=C(C=C1)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453917
|
Thermo Scientific Alfa Aesar
A18354.06 |
5 g |
44.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463917
|
Thermo Scientific Alfa Aesar
A18354.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
146949-07-7 | |
218.70 | |
LEFGAGRZHLNPLS-UHFFFAOYSA-N | |
2737222 | |
CCCC1=CC=C(C=C1)S(Cl)(=O)=O |
C9H11ClO2S | |
MFCD00041508 | |
4-n-propylbenzenesulfonyl chloride, 4-propylbenzene-1-sulfonyl chloride, 4-propyl-benzenesulfonylchloride, 4-propyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-propyl, p-propylbenzenesulfonyl chloride, 4-n-propylbenzenesulfonylchloride, acmc-209cxt, 4-propylphenylsulfonyl chloride, 4-propylphenylsulphonyl chloride | |
4-propylbenzenesulfonyl chloride |
Specifications
146949-07-7 | |
1.223 | |
C9H11ClO2S | |
MFCD00041508 | |
5511216 | |
4-n-propylbenzenesulfonyl chloride, 4-propylbenzene-1-sulfonyl chloride, 4-propyl-benzenesulfonylchloride, 4-propyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-propyl, p-propylbenzenesulfonyl chloride, 4-n-propylbenzenesulfonylchloride, acmc-209cxt, 4-propylphenylsulfonyl chloride, 4-propylphenylsulphonyl chloride | |
CCCC1=CC=C(C=C1)S(Cl)(=O)=O | |
218.70 | |
218.7 | |
4-n-Propylbenzenesulfonyl chloride |
20°C to 22°C | |
275°C to 276°C | |
1.5375 | |
UN3265 | |
Moisture sensitive | |
LEFGAGRZHLNPLS-UHFFFAOYSA-N | |
4-propylbenzenesulfonyl chloride | |
2737222 | |
95% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.