missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxypiperidine, 95%, Thermo Scientific Chemicals

Product Code. 11469740
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88536-11-2
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD06248727
InChI Key VOWMRECKIQVVPP-UHFFFAOYSA-N
Synonym 4-propoxy-piperidine, 4-n-propoxypiperidine, 4-propyloxypiperidine, 4-n-propoxypiperidin, acmc-20ambz, 4-piperidyl propyl ether, 4-piperidinyl propyl ether, piperidine, 4-propoxy-, hydrochloride
PubChem CID 11492037
IUPAC Name 4-propoxypiperidine
SMILES CCCOC1CCNCC1

Specifications

Quantity 1 g
Formula Weight 143.23
Percent Purity 95%
Chemical Name or Material 4-n-Propoxypiperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'