missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Propoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific™

35.78 € - 103.00 €

Chemical Identifiers

CAS 91062-38-3
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD06659852
InChI Key XJEONFAJOXSZDX-UHFFFAOYSA-N
Synonym 4-propoxycarbonyl phenylboronic acid, propyl 4-boronobenzoate, 4-n-propyloxycarbonylphenylboronic acid, n-propyl 4-boronobenzoate, 4-propoxycarbonylphenyl boronic acid, 4-propoxycarbonyl phenyl boronic acid, 4-n-propoxycarbonyl phenylboronic acid, 4-propoxycarbonyl benzene boronic acid, 4-n-propoxycarbonyl benzeneboronic acid
PubChem CID 23005364
IUPAC Name (4-propoxycarbonylphenyl)boronic acid
SMILES CCCOC(=O)C1=CC=C(C=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327539
View Documents
Thermo Scientific Alfa Aesar
H27938.03
1 g
35.78 €
1g
Please to purchase this item. Need a web account? Register with us today!
11337539
View Documents
Thermo Scientific Alfa Aesar
H27938.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

91062-38-3
208.02
XJEONFAJOXSZDX-UHFFFAOYSA-N
23005364
CCCOC(=O)C1=CC=C(C=C1)B(O)O
C10H13BO4
MFCD06659852
4-propoxycarbonyl phenylboronic acid, propyl 4-boronobenzoate, 4-n-propyloxycarbonylphenylboronic acid, n-propyl 4-boronobenzoate, 4-propoxycarbonylphenyl boronic acid, 4-propoxycarbonyl phenyl boronic acid, 4-n-propoxycarbonyl phenylboronic acid, 4-propoxycarbonyl benzene boronic acid, 4-n-propoxycarbonyl benzeneboronic acid
(4-propoxycarbonylphenyl)boronic acid

Specifications

91062-38-3
C10H13BO4
4-propoxycarbonyl phenylboronic acid, propyl 4-boronobenzoate, 4-n-propyloxycarbonylphenylboronic acid, n-propyl 4-boronobenzoate, 4-propoxycarbonylphenyl boronic acid, 4-propoxycarbonyl phenyl boronic acid, 4-n-propoxycarbonyl phenylboronic acid, 4-propoxycarbonyl benzene boronic acid, 4-n-propoxycarbonyl benzeneboronic acid
CCCOC(=O)C1=CC=C(C=C1)B(O)O
208.02
208.02
4-(n-Propoxycarbonyl)benzeneboronic acid
121°C to 123°C
MFCD06659852
XJEONFAJOXSZDX-UHFFFAOYSA-N
(4-propoxycarbonylphenyl)boronic acid
23005364
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.