missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylbiphenyl, 99%, Thermo Scientific™

Product Code. 11494304
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7116-96-3
Molecular Formula C17H20
Molecular Weight (g/mol) 224.35
MDL Number MFCD00009503
InChI Key IFUOTAQBVGAZPR-UHFFFAOYSA-N
Synonym 4-pentylbiphenyl, 4-n-pentylbiphenyl, 4-pentyl-1,1'-biphenyl, p-pentylbiphenyl, 1,1'-biphenyl, 4-pentyl, 4pentylbiphenyl, 1,1'-biphenyl, pentyl, 4-n-amyldiphenyl, 4-n-amylbiphenyl, pubchem12931
PubChem CID 81546
IUPAC Name 1-pentyl-4-phenylbenzene
SMILES CCCCCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 0.945
Boiling Point 166°C to 167°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.57
Quantity 5 g
Beilstein 2082056
Formula Weight 224.35
Percent Purity 99%
Chemical Name or Material 4-n-Pentylbiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'