Learn More
4-n-Pentylbenzoyl chloride, 98%, Thermo Scientific Chemicals
56.70 € - 741.00 €
Chemical Identifiers
CAS | 49763-65-7 |
---|---|
Molecular Formula | C12H15ClO |
Molecular Weight (g/mol) | 210.701 |
MDL Number | MFCD00000700 |
InChI Key | FBBRKYLXMNQFQU-UHFFFAOYSA-N |
Synonym | benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride |
PubChem CID | 170812 |
IUPAC Name | 4-pentylbenzoyl chloride |
SMILES | CCCCCC1=CC=C(C=C1)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433953
|
Thermo Scientific Alfa Aesar
A10248.06 |
5 g |
56.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443953
|
Thermo Scientific Alfa Aesar
A10248.14 |
25 g |
185.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453953
|
Thermo Scientific Alfa Aesar
A10248.22 |
100 g |
741.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
Chemical Identifiers
49763-65-7 | |
210.701 | |
FBBRKYLXMNQFQU-UHFFFAOYSA-N | |
170812 | |
CCCCCC1=CC=C(C=C1)C(=O)Cl |
C12H15ClO | |
MFCD00000700 | |
benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride | |
4-pentylbenzoyl chloride |
Specifications
49763-65-7 | |
144°C to 145°C (10 mmHg) | |
C12H15ClO | |
MFCD00000700 | |
2207887 | |
benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride | |
FBBRKYLXMNQFQU-UHFFFAOYSA-N | |
4-pentylbenzoyl chloride | |
170812 | |
98% |
1.034 | |
>110°C (230°F) | |
1.53 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CCCCCC1=CC=C(C=C1)C(=O)Cl | |
210.701 | |
210.71 | |
4-n-Pentylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 256-478-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only