missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptylphenol, 98+%, Thermo Scientific Chemicals

46.03 € - 453.26 €

Chemical Identifiers

CAS 1987-50-4
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00041751
InChI Key KNDDEFBFJLKPFE-UHFFFAOYSA-N
Synonym 4-n-heptylphenol, phenol, 4-heptyl, p-heptylphenol, heptanyl-p-phenol, p-hydroxyheptylbenzene, phenol, heptyl derivs., p-n-heptylphenol, unii-i743z35dvb, phenol, heptenylated, p-heptyl phenol
PubChem CID 16143
ChEBI CHEBI:34438
IUPAC Name 4-heptylphenol
SMILES CCCCCCCC1=CC=C(C=C1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478376
View Documents
Thermo Scientific Alfa Aesar
A15407.03
1 g
46.03 €
1g
Please to purchase this item. Need a web account? Register with us today!
11488376
View Documents
Thermo Scientific Alfa Aesar
A15407.06
5 g
162.81 €
5g
Please to purchase this item. Need a web account? Register with us today!
11498376
View Documents
Thermo Scientific Alfa Aesar
A15407.14
25 g
453.26 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1987-50-4
192.302
KNDDEFBFJLKPFE-UHFFFAOYSA-N
16143
4-heptylphenol
C13H20O
MFCD00041751
4-n-heptylphenol, phenol, 4-heptyl, p-heptylphenol, heptanyl-p-phenol, p-hydroxyheptylbenzene, phenol, heptyl derivs., p-n-heptylphenol, unii-i743z35dvb, phenol, heptenylated, p-heptyl phenol
CHEBI:34438
CCCCCCCC1=CC=C(C=C1)O

Specifications

1987-50-4
156°C (9 mmHg)
1.507
UN2430
KNDDEFBFJLKPFE-UHFFFAOYSA-N
4-heptylphenol
16143
192.3
4-n-Heptylphenol
25°C to 28°C
C13H20O
MFCD00041751
4-n-heptylphenol, phenol, 4-heptyl, p-heptylphenol, heptanyl-p-phenol, p-hydroxyheptylbenzene, phenol, heptyl derivs., p-n-heptylphenol, unii-i743z35dvb, phenol, heptenylated, p-heptyl phenol
CCCCCCCC1=CC=C(C=C1)O
192.302
CHEBI:34438
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 217-862-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.