Learn More
4'-(Methylthio)acetophenone, 99%, Thermo Scientific Chemicals
83.30 € - 295.00 €
Chemical Identifiers
CAS | 1778-09-2 |
---|---|
Molecular Formula | C9H10OS |
Molecular Weight (g/mol) | 166.24 |
MDL Number | MFCD00039835 |
InChI Key | JECUZQLBQKNEMW-UHFFFAOYSA-N |
Synonym | 4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone |
PubChem CID | 74501 |
IUPAC Name | 1-(4-methylsulfanylphenyl)ethanone |
SMILES | CSC1=CC=C(C=C1)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334905
|
Thermo Scientific Alfa Aesar
L18074.06 |
5 g |
83.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344905
|
Thermo Scientific Alfa Aesar
L18074.14 |
25 g |
295.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1778-09-2 | |
166.24 | |
JECUZQLBQKNEMW-UHFFFAOYSA-N | |
74501 | |
CSC1=CC=C(C=C1)C(C)=O |
C9H10OS | |
MFCD00039835 | |
4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone | |
1-(4-methylsulfanylphenyl)ethanone |
Specifications
1778-09-2 | |
135°C (1 mmHg) | |
MFCD00039835 | |
4'-methylthio acetophenone, 4-methylthio acetophenone, 4'-methylthioacetophenone, 1-4-methylsulfanyl phenyl ethanone, 4-acetyl thioanisole, 4'-methylmercapto acetophenone, 1-4-methylthio phenyl ethanone, 1-4-methylthio phenyl ethan-1-one, 1-4-methylsulfanylphenyl ethanone | |
CSC1=CC=C(C=C1)C(C)=O | |
166.24 | |
166.24 | |
4'-(Methylthio)acetophenone |
80°C to 82°C | |
C9H10OS | |
1860517 | |
JECUZQLBQKNEMW-UHFFFAOYSA-N | |
1-(4-methylsulfanylphenyl)ethanone | |
74501 | |
99% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 217-213-1
missing translation for 'tsca' : No
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.