missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methylpropiophenone, 94%, Thermo Scientific Chemicals
45.10 € - 175.00 €
Chemical Identifiers
CAS | 5337-93-9 |
---|---|
Molecular Formula | C10H12O |
Molecular Weight (g/mol) | 148.21 |
MDL Number | MFCD00009312 |
InChI Key | PATYHUUYADUHQS-UHFFFAOYSA-N |
Synonym | 4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone |
PubChem CID | 21429 |
ChEBI | CHEBI:59335 |
IUPAC Name | 1-(4-methylphenyl)propan-1-one |
SMILES | CCC(=O)C1=CC=C(C)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420257
|
Thermo Scientific Alfa Aesar
B21673.22 |
100 g |
45.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430257
|
Thermo Scientific Alfa Aesar
B21673.36 |
500 g |
175.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4?-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.
Solubility
Soluble in chloroform and hexane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
5337-93-9 | |
148.21 | |
PATYHUUYADUHQS-UHFFFAOYSA-N | |
21429 | |
1-(4-methylphenyl)propan-1-one |
C10H12O | |
MFCD00009312 | |
4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone | |
CHEBI:59335 | |
CCC(=O)C1=CC=C(C)C=C1 |
Specifications
5337-93-9 | |
0.991 | |
108°C (226°F) | |
1.528 | |
2042137 | |
Soluble in chloroform and hexane. | |
CCC(=O)C1=CC=C(C)C=C1 | |
148.21 | |
CHEBI:59335 | |
94% |
7°C | |
238°C to 239°C | |
C10H12O | |
MFCD00009312 | |
4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone | |
PATYHUUYADUHQS-UHFFFAOYSA-N | |
1-(4-methylphenyl)propan-1-one | |
21429 | |
148.21 | |
4'-Methylpropiophenone |
Safety and Handling
missing translation for 'einecsNumber' : 226-267-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only