Learn More
4-Methylpiperidine, 99%, Thermo Scientific Chemicals
31.70 € - 116.00 €
Chemical Identifiers
CAS | 626-58-4 |
---|---|
Molecular Formula | C6H13N |
Molecular Weight (g/mol) | 99.18 |
MDL Number | MFCD00006005 |
InChI Key | UZOFELREXGAFOI-UHFFFAOYSA-N |
Synonym | 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline |
PubChem CID | 69381 |
IUPAC Name | 4-methylpiperidine |
SMILES | CC1CCNCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10123180
|
Thermo Scientific Acros
127511000 |
100 g |
31.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10487130
|
Thermo Scientific Acros
127515000 |
500 g |
116.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
626-58-4 | |
99.18 | |
UZOFELREXGAFOI-UHFFFAOYSA-N | |
69381 | |
CC1CCNCC1 |
C6H13N | |
MFCD00006005 | |
4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline | |
4-methylpiperidine |
Specifications
626-58-4 | |
100.0 | |
Yellow | |
122.0°C | |
Authentic | |
Glass bottle | |
1.4448 to 1.4468 | |
0.84 | |
Solubility in water: miscible. | |
CC1CCNCC1 | |
99.18 | |
99.18 | |
Liquid |
98.5 | |
4.0°C to 5.0°C | |
0.8400g/mL | |
13°C | |
98.5% min. (GC) | |
C6H13N | |
MFCD00006005 | |
4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline | |
UZOFELREXGAFOI-UHFFFAOYSA-N | |
4-methylpiperidine | |
69381 | |
99% | |
4-Methylpiperidine, 99% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-954-1
missing translation for 'tsca' : TSCA
- 4-Methylpiperidine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Flammable liquid Category 2
Hazard Statement
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.,IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.,IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/physician.,Immediately call a POISON CENTER/doctor/physician.
RUO – Research Use Only