missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific Chemicals

Product Code. 11325335
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380481-66-3
Molecular Formula C12H17BO2
Molecular Weight (g/mol) 204.076
MDL Number MFCD03788728
InChI Key ZSPBWRPQSPRISS-UHFFFAOYSA-N
Synonym 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane
PubChem CID 23005406
IUPAC Name 5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C

Specifications

Melting Point 93°C to 95°C
Quantity 25 g
Formula Weight 204.08
Percent Purity 99%
Chemical Name or Material 4-Methylbenzeneboronic acid neopentyl glycol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'