Learn More
4-Methylbenzamidoxime, 97%, Thermo Scientific™
59.00 € - 238.19 €
Chemical Identifiers
CAS | 19227-13-5 |
---|---|
Molecular Formula | C8H10N2O |
Molecular Weight (g/mol) | 150.181 |
MDL Number | MFCD00019952 |
InChI Key | NKJXMLIWSJATEE-UHFFFAOYSA-N |
Synonym | 4-methylbenzamidoxime, 4-methylbenzamide oxime, p-toluamidoxime, n-hydroxy-4-methyl-benzamidine, hydroxyimino 4-methylphenyl methylamine, n'-hydroxy-4-methylbenzene-1-carboximidamide, e-n'-hydroxy-4-methylbenzenecarboximidamide, z-n'-hydroxy-4-methylbenzene-1-carboximidamide, n'-hydroxy-4-methylbenzimidamide, n-hydroxy-4-methylbenzenecarboximidamide |
PubChem CID | 5493168 |
IUPAC Name | N'-hydroxy-4-methylbenzenecarboximidamide |
SMILES | CC1=CC=C(C=C1)C(=NO)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428157
|
Thermo Scientific Alfa Aesar
H51704.03 |
1 g | N/A | ||||||
15438157
|
Thermo Scientific Alfa Aesar
H51704.06 |
5 g |
238.19 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
19227-13-5 | |
150.181 | |
NKJXMLIWSJATEE-UHFFFAOYSA-N | |
5493168 | |
CC1=CC=C(C=C1)C(=NO)N |
C8H10N2O | |
MFCD00019952 | |
4-methylbenzamidoxime, 4-methylbenzamide oxime, p-toluamidoxime, n-hydroxy-4-methyl-benzamidine, hydroxyimino 4-methylphenyl methylamine, n'-hydroxy-4-methylbenzene-1-carboximidamide, e-n'-hydroxy-4-methylbenzenecarboximidamide, z-n'-hydroxy-4-methylbenzene-1-carboximidamide, n'-hydroxy-4-methylbenzimidamide, n-hydroxy-4-methylbenzenecarboximidamide | |
N'-hydroxy-4-methylbenzenecarboximidamide |
Specifications
19227-13-5 | |
C8H10N2O | |
4-methylbenzamidoxime, 4-methylbenzamide oxime, p-toluamidoxime, n-hydroxy-4-methyl-benzamidine, hydroxyimino 4-methylphenyl methylamine, n'-hydroxy-4-methylbenzene-1-carboximidamide, e-n'-hydroxy-4-methylbenzenecarboximidamide, z-n'-hydroxy-4-methylbenzene-1-carboximidamide, n'-hydroxy-4-methylbenzimidamide, n-hydroxy-4-methylbenzenecarboximidamide | |
CC1=CC=C(C=C1)C(=NO)N | |
150.181 | |
150.18 | |
4-Methylbenzamidoxime |
135°C to 138°C | |
MFCD00019952 | |
NKJXMLIWSJATEE-UHFFFAOYSA-N | |
N'-hydroxy-4-methylbenzenecarboximidamide | |
5493168 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'rtecsNumber' : XS4870000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.