missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, 96%, Thermo Scientific Chemicals

Product Code. 11461788
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C)C=C1

Specifications

Melting Point -19°C
Density 1.01
Boiling Point 224°C to 226°C
Flash Point 92°C (197°F)
Refractive Index 1.534
Quantity 1000 g
Beilstein 606053
Solubility Information Miscible with water.
Formula Weight 134.18
Percent Purity 96%
Chemical Name or Material 4'-Methylacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'