missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, Honeywell
105.85 € - 454.17 €
Chemical Identifiers
CAS | 108-10-1 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.161 |
MDL Number | MFCD00008938 |
InChI Key | NTIZESTWPVYFNL-UHFFFAOYSA-N |
Synonym | 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon |
PubChem CID | 7909 |
ChEBI | CHEBI:82344 |
IUPAC Name | 4-methylpentan-2-one |
SMILES | CC(C)CC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15623980
|
Honeywell Riedel-de Haën
33463-1L |
1 L |
105.85 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15633980
|
Honeywell Riedel-de Haën
33463-2.5L |
2.5 L |
221.35 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643980
|
Honeywell Riedel-de Haën
33463-20KG |
20 kg | N/A | ||||||
Description
Chemical Identifiers
108-10-1 | |
100.161 | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
7909 | |
4-methylpentan-2-one |
C6H12O | |
MFCD00008938 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CHEBI:82344 | |
CC(C)CC(=O)C |
Specifications
108-10-1 | |
14°C (57.2°F) | |
(CH3)2CHCH2COCH3 | |
UN1245 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CC(C)CC(=O)C | |
100.161 | |
3.5 (vs air) | |
100.16g/mol | |
4-Methyl-2-pentanone |
0.801g/mL at 25°C | |
C6H12O | |
MFCD00008938 | |
605399 | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
7909 | |
CHEBI:82344 | |
≥99% |
Safety and Handling
P210-P261-P305 + P351 + P338
Danger
missing translation for 'einecsNumber' : 203-550-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 4-Methyl-2-pentanone
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 4-methylpentan-2-one
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title