Learn More
4-Methyl-1-pentyne, 97%, Thermo Scientific Chemicals
51.50 € - 161.00 €
Chemical Identifiers
CAS | 7154-75-8 |
---|---|
Molecular Formula | C6H10 |
Molecular Weight (g/mol) | 82.15 |
MDL Number | MFCD00039857 |
InChI Key | OXRWICUICBZVAE-UHFFFAOYSA-N |
Synonym | 4-methyl-1-pentyne, 1-pentyne, 4-methyl, isobutylacetylene, isobutylethyne, 4-methylpentine, 4-methylpentyne, 4-methyl pentyne, isobutyl acetylene, 4-methyl-pentyne, 4-methyl-pent-1-yne |
PubChem CID | 138948 |
IUPAC Name | 4-methylpent-1-yne |
SMILES | CC(C)CC#C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357485
|
Thermo Scientific Alfa Aesar
L05537.03 |
1 g |
51.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367485
|
Thermo Scientific Alfa Aesar
L05537.06 |
5 g |
161.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.
Solubility
Insoluble in water. Soluble in common organic solvents of low polarity.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
7154-75-8 | |
82.15 | |
OXRWICUICBZVAE-UHFFFAOYSA-N | |
138948 | |
CC(C)CC#C |
C6H10 | |
MFCD00039857 | |
4-methyl-1-pentyne, 1-pentyne, 4-methyl, isobutylacetylene, isobutylethyne, 4-methylpentine, 4-methylpentyne, 4-methyl pentyne, isobutyl acetylene, 4-methyl-pentyne, 4-methyl-pent-1-yne | |
4-methylpent-1-yne |
Specifications
7154-75-8 | |
61°C to 62°C | |
C6H10 | |
MFCD00039857 | |
1731077 | |
Insoluble in water. Soluble in common organic solvents of low polarity. | |
CC(C)CC#C | |
82.15 | |
82.15 | |
4-Methyl-1-pentyne |
0.698 | |
−24°C (−11°F) | |
1.393 | |
UN3295 | |
4-methyl-1-pentyne, 1-pentyne, 4-methyl, isobutylacetylene, isobutylethyne, 4-methylpentine, 4-methylpentyne, 4-methyl pentyne, isobutyl acetylene, 4-methyl-pentyne, 4-methyl-pent-1-yne | |
OXRWICUICBZVAE-UHFFFAOYSA-N | |
4-methylpent-1-yne | |
138948 | |
97% |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only