missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthoyl chloride, 98%, Thermo Scientific™

Product Code. 15442339
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87700-67-2
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.653
MDL Number MFCD12025362
InChI Key IXGMHYHWSPDNNS-UHFFFAOYSA-N
Synonym 1-naphthalenecarbonylchloride, 4-methyl, 1-naphthalenecarbonyl chloride, 4-methyl, 4-methyl-1-naphthalenecarbonyl chloride, 1-naphthalenecarbonyl chloride, 4-methyl-9ci, 4-methyl-1-napthoyl chloride, 4-methyl-1-naphthoyl chloride, 4-methyl-naphthalene-1-carbonyl chloride
PubChem CID 24887412
IUPAC Name 4-methylnaphthalene-1-carbonyl chloride
SMILES CC1=CC=C(C2=CC=CC=C12)C(=O)Cl

Specifications

Color Yellow
Quantity 5 g
UN Number UN3261
Beilstein 3247818
Formula Weight 204.65
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Methyl-1-naphthoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.