missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10396974
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 18212-21-0
Molecular Formula C4H3N2O2S
Molecular Weight (g/mol) 143.14
MDL Number MFCD00173830
InChI Key NHHQOYLPBUYHQU-UHFFFAOYSA-M
Synonym 4-methyl-1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl, pubchem10092, acmc-209xfo, 4-methyl-1,2,3-thiadiazole-5-carboxylicacid, tos-bb-0050, 4-methyl-1,2,3 thiadiazol-5-carboxylic acid, 4-methyl-5-hydroxycarbonyl-1,2,3-thiadiazole, 4methyl-1,2,3 thiadiazole-5-carboxylic acid
PubChem CID 1381992
IUPAC Name 4-methylthiadiazole-5-carboxylic acid
SMILES CC1=C(SN=N1)C([O-])=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 144.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'