missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxypropiophenone, 99%, Thermo Scientific Chemicals

49.85 € - 513.56 €

Chemical Identifiers

CAS 121-97-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009310
InChI Key ZJVAWPKTWVFKHG-UHFFFAOYSA-N
PubChem CID 67144
IUPAC Name 1-(4-methoxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11417566
View Documents
Thermo Scientific Alfa Aesar
A14619.14
25 g
49.85 €
25g
Please to purchase this item. Need a web account? Register with us today!
11427566
View Documents
Thermo Scientific Alfa Aesar
A14619.22
100 g
132.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
11437566
View Documents
Thermo Scientific Alfa Aesar
A14619.36
500 g
513.56 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-97-1
164.204
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
1-(4-methoxyphenyl)propan-1-one
C10H12O2
MFCD00009310
67144
CCC(=O)C1=CC=C(C=C1)OC

Specifications

121-97-1
1.07
62°C (143°F)
1.5465
907733
ZJVAWPKTWVFKHG-UHFFFAOYSA-N
1-(4-methoxyphenyl)propan-1-one
67144
99%
27°C to 29°C
273°C to 275°C
C10H12O2
MFCD00009310
4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
CCC(=O)C1=CC=C(C=C1)OC
164.204
164.2
4'-Methoxypropiophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H332-H315-H319-H335

missing translation for 'einecsNumber' : 204-512-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only