missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxypiperidine, 98+%, Thermo Scientific Chemicals

128.64 € - 407.03 €

Chemical Identifiers

CAS 4045-24-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD04115010
InChI Key ZEYSHALLPAKUHG-UHFFFAOYSA-N
Synonym 4-methoxy-piperidine, piperidine, 4-methoxy, hcl, 4-methoxypiperdine, 4-methoxy piperidine, 4-methoxyl piperidine, pubchem8024
PubChem CID 77666
IUPAC Name 4-methoxypiperidine
SMILES COC1CCNCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10402785
View Documents
Thermo Scientific Acros
393390010
1 g Glass bottle
128.64 €
1g
Please to purchase this item. Need a web account? Register with us today!
10248753
View Documents
Thermo Scientific Acros
393390050
5 g Glass bottle
407.03 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4045-24-3
115.18
ZEYSHALLPAKUHG-UHFFFAOYSA-N
77666
COC1CCNCC1
C6H13NO
MFCD04115010
4-methoxy-piperidine, piperidine, 4-methoxy, hcl, 4-methoxypiperdine, 4-methoxy piperidine, 4-methoxyl piperidine, pubchem8024
4-methoxypiperidine

Specifications

4045-24-3
100.0
44.0°C (9.0 Mbar)
98% min. (GC)
1.4590 to 1.4610 (20°C, 589nm)
21, V,1, 51
4-methoxy-piperidine, piperidine, 4-methoxy, hcl, 4-methoxypiperdine, 4-methoxy piperidine, 4-methoxyl piperidine, pubchem8024
COC1CCNCC1
115.18
115.18
Liquid
98.0
0.9300g/mL
Authentic
C6H13NO
MFCD04115010
0.93
ZEYSHALLPAKUHG-UHFFFAOYSA-N
4-methoxypiperidine
77666
98+%
4-Methoxypiperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 223-742-9

Product Identifier
  • 4-Methoxypiperidine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.