Learn More
4-Methoxyphenol, 98+%, Thermo Scientific Chemicals
20.30 € - 112.56 €
Chemical Identifiers
CAS | 150-76-5 |
---|---|
Molecular Formula | C7H8O2 |
Molecular Weight (g/mol) | 124.14 |
MDL Number | MFCD00002332 |
InChI Key | NWVVVBRKAWDGAB-UHFFFAOYSA-N |
Synonym | mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum |
PubChem CID | 9015 |
ChEBI | CHEBI:69441 |
IUPAC Name | 4-methoxyphenol |
SMILES | COC1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11472617
|
Thermo Scientific Alfa Aesar
A14845.18 |
50 g |
20.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11482617
|
Thermo Scientific Alfa Aesar
A14845.30 |
250 g |
40.40 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11462617
|
Thermo Scientific Alfa Aesar
A14845.0B |
1000 g |
112.56 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.
Solubility
Soluble in acetone, ethyl acetate, ethanol, ether, benzene and carbon tetrachloride.
Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with halogens and oxidizing agents.
Chemical Identifiers
150-76-5 | |
124.14 | |
NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
9015 | |
4-methoxyphenol |
C7H8O2 | |
MFCD00002332 | |
mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
CHEBI:69441 | |
COC1=CC=C(O)C=C1 |
Specifications
150-76-5 | |
1.55 | |
132°C (269°F) | |
MFCD00002332 | |
mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
4-methoxyphenol | |
9015 | |
124.14 | |
4-Methoxyphenol |
55°C to 58°C | |
243°C to 246°C | |
C7H8O2 | |
507924 | |
Soluble in acetone,ethyl acetate,ethanol,ether,benzene and carbon tetrachloride. | |
COC1=CC=C(O)C=C1 | |
124.14 | |
CHEBI:69441 | |
≥98% |
Safety and Handling
GHS H Statement
H302-H319-H317
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P363-P501c
H302-H317-H319-H361d-H373-H500
missing translation for 'einecsNumber' : 205-769-8
missing translation for 'rtecsNumber' : SL7700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.