missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Methoxycarbamoyl)benzeneboronic acid, 97%

Product Code. 15499947
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 850568-17-1
Molecular Formula C8H10BNO4
Molecular Weight (g/mol) 194.981
MDL Number MFCD04115670
InChI Key MZZWCLBDQROHHS-UHFFFAOYSA-N
Synonym 4-o-methylhydroxylaminocarbonyl phenylboronic acid, 4-methoxycarbamoyl phenyl boronic acid, 4-o-methylhydroxylaminocarbonyl benzeneboronic acid, 4-methoxycarbamoyl phenylboronic acid, 4-methoxycarbamoyl benzeneboronic acid, 4-o-methyl hydroxylaminocarbonyl phenylboronic acid, 4-o-methylhydroaminocarbonyl phenylboronic acid
PubChem CID 3379984
IUPAC Name [4-(methoxycarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NOC)(O)O

Specifications

Melting Point 130°C to 136°C
Quantity 250 mg
Formula Weight 194.98
Percent Purity 97%
Chemical Name or Material 4-(Methoxycarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.