missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10193234
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown to Yellow
CAS 38769-92-5, 109-99-9
CAS Min % 2.97, 93.07
CAS Max % 6.93, 97.03
Molecular Formula C8H9ClMgO
Linear Formula CH3OC6H4CH2MgCl
MDL Number MFCD01311491
Concentration 0.15 to 0.35M
Density 0.9140g/mL
Synonym 4-methoxybenzylmagnesium chloride, 4-methoxybenzylmagnesium chloride solution, magnesium, chloro 4-methoxyphenyl methyl, p-methoxybenzylmagnesium chloride, 4-methoxy benzylmagnesium chloride, xhdyucuvuzsism-uhfffaoysa-m, 4-methoxybenzyl magnesium chloride, chloro 4-methoxyphenyl methyl magnesium, 4-methoxybenzylmagnesium chloride 0.25 m in tetrahydrofuran, 4-methoxybenzylmagnesium chloride solution, 0.25 m in thf
InChI Key XHDYUCUVUZSISM-UHFFFAOYSA-M
SMILES COC1=CC=C(C[Mg]Cl)C=C1
IUPAC Name chloro[(4-methoxyphenyl)methyl]magnesium
Molecular Weight (g/mol) 180.91
PubChem CID 10965134
Formula Weight 180.91
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.914
Quantity 100 mL
Flash Point −17°C
Physical Form Solution
Chemical Name or Material 4-Methoxybenzylmagnesium chloride, 0.25M solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methoxybenzylmagnesium chloride, 0.25M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'