missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylcyclohexanone, 96%, Thermo Scientific Chemicals

Product Code. 11302766
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Isopropylcyclohexanone is used to prepare 4-isopropyl-cyclohexanone oxime. It is also used in the synthesis of dihydroindol-2-ones as agonists and antagonist ligands at the nociceptin receptor. Further, it is used in the preparation of beta-alanine derivatives, which acts as orally available glucagon receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylcyclohexanone is used to prepare 4-isopropyl-cyclohexanone oxime. It is also used in the synthesis of dihydroindol-2-ones as agonists and antagonist ligands at the nociceptin receptor. Further, it is used in the preparation of beta-alanine derivatives, which acts as orally available glucagon receptor antagonists.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with oxidizing agents. reducing agents and plastics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5432-85-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00043479
InChI Key FPKISACHVIIMRA-UHFFFAOYSA-N
Synonym 4-isopropylcyclohexanone, cyclohexanone, 4-1-methylethyl, 4-propan-2-yl cyclohexan-1-one, 4-isopropylcyclohexan-1-one, cyclohexanone, 4-isopropyl, 4-isopropyl-cyclohexanone, 4-iso-propylcyclohexanone, 4-methylethyl cyclohexan-1-one, acmc-20a5fk, 4-isopropyl cyclohexanone
PubChem CID 79488
IUPAC Name 4-propan-2-ylcyclohexan-1-one
SMILES CC(C)C1CCC(=O)CC1

Specifications

Boiling Point 94°C to 96°C (17 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.458
Quantity 10 g
Beilstein 1236952
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 4-Isopropylcyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.