missing translation for 'onlineSavingsMsg'
Learn More

4-(Isopropoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™

128.00 € - 545.00 €

Chemical Identifiers

CAS 342002-82-8
Molecular Formula C10H13BO4
Molecular Weight (g/mol) 208.02
MDL Number MFCD02259385
InChI Key NIQHFKWRHJZMQU-UHFFFAOYSA-N
Synonym 4-isopropoxycarbonylphenylboronic acid, 4-isopropoxycarbonyl phenyl boronic acid, 4-isopropoxycarbonyl benzeneboronic acid, propan-2-yl 4-dihydroxyboranyl benzoate, isopropyl 4-boronobenzoate, 4-isopropoxycarbonyl phenylboronic acid, p-isopropoxycarbonyl phenylboronic acid, 4-isopropoxycarbonylphenylboronicacid, 4-propan-2-yloxycarbonylphenyl boronic acid, acmc-209i5o
PubChem CID 2734703
IUPAC Name (4-propan-2-yloxycarbonylphenyl)boronic acid
SMILES CC(C)OC(=O)C1=CC=C(C=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15450078
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53335.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15460078
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H53335.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(Isopropoxycarbonyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

342002-82-8
208.02
NIQHFKWRHJZMQU-UHFFFAOYSA-N
2734703
CC(C)OC(=O)C1=CC=C(C=C1)B(O)O
C10H13BO4
MFCD02259385
4-isopropoxycarbonylphenylboronic acid, 4-isopropoxycarbonyl phenyl boronic acid, 4-isopropoxycarbonyl benzeneboronic acid, propan-2-yl 4-dihydroxyboranyl benzoate, isopropyl 4-boronobenzoate, 4-isopropoxycarbonyl phenylboronic acid, p-isopropoxycarbonyl phenylboronic acid, 4-isopropoxycarbonylphenylboronicacid, 4-propan-2-yloxycarbonylphenyl boronic acid, acmc-209i5o
(4-propan-2-yloxycarbonylphenyl)boronic acid

Specifications

342002-82-8
C10H13BO4
4-isopropoxycarbonylphenylboronic acid, 4-isopropoxycarbonyl phenyl boronic acid, 4-isopropoxycarbonyl benzeneboronic acid, propan-2-yl 4-dihydroxyboranyl benzoate, isopropyl 4-boronobenzoate, 4-isopropoxycarbonyl phenylboronic acid, p-isopropoxycarbonyl phenylboronic acid, 4-isopropoxycarbonylphenylboronicacid, 4-propan-2-yloxycarbonylphenyl boronic acid, acmc-209i5o
CC(C)OC(=O)C1=CC=C(C=C1)B(O)O
208.02
208.02
4-(Isopropoxycarbonyl)benzeneboronic acid
111°C
MFCD02259385
NIQHFKWRHJZMQU-UHFFFAOYSA-N
(4-propan-2-yloxycarbonylphenyl)boronic acid
2734703
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications