missing translation for 'onlineSavingsMsg'
Learn More

4-Iodopyridine, ≥97%, Thermo Scientific™

25.80 € - 367.00 €

Chemical Identifiers

CAS 15854-87-2
Molecular Formula C5H4IN
Molecular Weight (g/mol) 205.00
MDL Number MFCD02093937
InChI Key RTLUPHDWSUGAOS-UHFFFAOYSA-N
Synonym pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
PubChem CID 609492
IUPAC Name 4-iodopyridine
SMILES IC1=CC=NC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10462723
View Documents
Thermo Scientific Maybridge
MO00014DA
1 g
25.80 €
1g
Please to purchase this item. Need a web account? Register with us today!
10055322
View Documents
Thermo Scientific Maybridge
MO00014EA
10 g
195.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10515835
View Documents
Thermo Scientific Maybridge
MO00014EB
25 g
367.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

15854-87-2
205.00
RTLUPHDWSUGAOS-UHFFFAOYSA-N
609492
IC1=CC=NC=C1
C5H4IN
MFCD02093937
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
4-iodopyridine

Specifications

15854-87-2
97%
C5H4IN
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d
IC1=CC=NC=C1
205.00
204.99
4-Iodopyridine
97.0
Amber glass bottle
MFCD02093937
RTLUPHDWSUGAOS-UHFFFAOYSA-N
4-iodopyridine
609492
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Freezer -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.