Learn More
4-Iodopyridine, 96%, Thermo Scientific Chemicals
42.11 € - 173.87 €
Chemical Identifiers
CAS | 15854-87-2 |
---|---|
Molecular Formula | C5H4IN |
Molecular Weight (g/mol) | 205.00 |
MDL Number | MFCD02093937 |
InChI Key | RTLUPHDWSUGAOS-UHFFFAOYSA-N |
Synonym | pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d |
PubChem CID | 609492 |
IUPAC Name | 4-iodopyridine |
SMILES | IC1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10238943
|
Thermo Scientific Acros
|
1 g | Glass bottle |
42.11 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10550554
|
Thermo Scientific Acros
|
5 g | Glass bottle |
173.87 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15854-87-2 | |
205.00 | |
RTLUPHDWSUGAOS-UHFFFAOYSA-N | |
609492 | |
IC1=CC=NC=C1 |
C5H4IN | |
MFCD02093937 | |
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d | |
4-iodopyridine |
Specifications
15854-87-2 | |
100.0 | |
Brown to Brown-Green | |
95% min. (GC) | |
MFCD02093937 | |
Solubility in water: slightly soluble | |
IC1=CC=NC=C1 | |
205.00 | |
204.99 | |
Crystals or Powder |
95.0 | |
92.0°C to 96.0°C | |
Authentic | |
C5H4IN | |
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d | |
RTLUPHDWSUGAOS-UHFFFAOYSA-N | |
4-iodopyridine | |
609492 | |
96% | |
4-Iodopyridine |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
Recommended Storage : Product darkens on storage
- 4-Iodopyridine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.