Learn More
4'-Iodoacetophenone, 98%, Thermo Scientific Chemicals
34.40 € - 112.00 €
Chemical Identifiers
CAS | 13329-40-3 |
---|---|
Molecular Formula | C8H7IO |
Molecular Weight (g/mol) | 246.05 |
MDL Number | MFCD00045320 |
InChI Key | JZJWCDQGIPQBAO-UHFFFAOYSA-N |
Synonym | 4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone |
PubChem CID | 72869 |
IUPAC Name | 1-(4-iodophenyl)ethanone |
SMILES | CC(=O)C1=CC=C(C=C1)I |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10415755
|
Thermo Scientific Acros
278890050 |
5 g | Glass bottle |
34.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10470141
|
Thermo Scientific Acros
278890250 |
25 g | Glass bottle |
112.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13329-40-3 | |
246.05 | |
JZJWCDQGIPQBAO-UHFFFAOYSA-N | |
72869 | |
CC(=O)C1=CC=C(C=C1)I |
C8H7IO | |
MFCD00045320 | |
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone | |
1-(4-iodophenyl)ethanone |
Specifications
13329-40-3 | |
100.0 | |
Brown | |
97.5% min. (GC) | |
IC6H4C(O)CH3 | |
4'-iodoacetophenone, 1-4-iodophenyl ethanone, 4-iodoacetophenone, 1-4-iodophenyl ethan-1-one, p-iodoacetophenone, ethanone, 1-4-iodophenyl, 1-4-iodo-phenyl-ethanone, 1-acetyl-4-iodobenzene, 4-iodo acetophenone, 4;-iodoacetophenone | |
CC(=O)C1=CC=C(C=C1)I | |
246.05 | |
246.05 | |
Crystalline Powder |
97.5 | |
82.0°C to 89.0°C | |
Authentic | |
C8H7IO | |
MFCD00045320 | |
JZJWCDQGIPQBAO-UHFFFAOYSA-N | |
1-(4-iodophenyl)ethanone | |
72869 | |
98% | |
4'-Iodoacetophenone, 98% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-372-8
missing translation for 'tsca' : TSCA
- 4'-Iodoacetophenone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only