missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-2-methyl-1H-imidazole, 97%, Thermo Scientific™
158.00 € - 555.00 €
Chemical Identifiers
CAS | 73746-45-9 |
---|---|
Molecular Formula | C4H5IN2 |
Molecular Weight (g/mol) | 208.00 |
MDL Number | MFCD02684335 |
InChI Key | SEDSLMWYUQACGR-UHFFFAOYSA-N |
PubChem CID | 2794623 |
IUPAC Name | 5-iodo-2-methyl-1H-imidazole |
SMILES | CC1=NC=C(I)N1 |
Description
Chemical Identifiers
73746-45-9 | |
208.00 | |
SEDSLMWYUQACGR-UHFFFAOYSA-N | |
5-iodo-2-methyl-1H-imidazole |
C4H5IN2 | |
MFCD02684335 | |
2794623 | |
CC1=NC=C(I)N1 |
Specifications
73746-45-9 | |
Authentic | |
C4H5IN2 | |
4-iodo-2-methyl-1h-imidazole, 2-methyl-4 5-iodo-1 h-imidazole, 4-iodo-2-methylimidazole, 2-methyl-4-iodo-1h-imidazole, 4-iodo-2-methyl-3h-imidazole, 1h-imidazole, 4-iodo-2-methyl, 2-methyl-4-jodimidazol, ksc638k7d, 1h-imidazole,4-iodo-2-methyl, 1h-imidazole,5-iodo-2-methyl | |
CC1=NC=C(I)N1 | |
208.00 | |
208 | |
4-Iodo-2-methyl-1H-imidazole |
138.0°C to 144.0°C | |
96% min. (HPLC) | |
MFCD02684335 | |
SEDSLMWYUQACGR-UHFFFAOYSA-N | |
5-iodo-2-methyl-1H-imidazole | |
2794623 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Iodo-2-methyl-1H-imidazole, 97%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title