missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1-benzyl-1H-pyrazole, 95%, Thermo Scientific™

Product Code. 15487397
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50877-42-4
Molecular Formula C10H9IN2
Molecular Weight (g/mol) 284.1
MDL Number MFCD04038745
InChI Key PVEYRBGIYMWFPB-UHFFFAOYSA-N
Synonym 1-benzyl-4-iodo-1h-pyrazole, 1h-pyrazole, 4-iodo-1-phenylmethyl, benzyl-4-iodo-1h-pyrazole, 1h-n-benzyl-4-iodopyrazole, 1-benzyl-4-iodo-pyrazole, zlchem 893, pubchem23732, acmc-209kqm, n-benzyl-4-iodopyrazole, 4-iodo-1-benzylpyrazole
PubChem CID 3794579
IUPAC Name 1-benzyl-4-iodopyrazole
SMILES C1=CC=C(C=C1)CN2C=C(C=N2)I

Specifications

Quantity 5 g
Formula Weight 284.1
Percent Purity 95%
Chemical Name or Material 4-Iodo-1-benzyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'