missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxythiobenzamide, 98%, Thermo Scientific Chemicals

92.20 € - 298.00 €

Chemical Identifiers

CAS 25984-63-8
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.20
MDL Number MFCD04973332
InChI Key DOCQBKMMNPJLOR-UHFFFAOYSA-N
Synonym 4-hydroxythiobenzamide, 4-hydroxybenzothioamide, 4-hydroxy thiobenzamide, 4-hydroxybenzene-1-carbothioamide, 4-hydroxybenzenecarbothioamide, benzenecarbothioamide, 4-hydroxy, 4-hydroxy-thiobenzamide, 4-aminothioxomethyl phenol, 4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one, p-hydroxythiobenzamide
PubChem CID 5706487
IUPAC Name 4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES NC(S)=C1C=CC(=O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32780.06
5 g
92.20 €
5g
Please to purchase this item. Need a web account? Register with us today!
15475537
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32780.14
25 g
298.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25984-63-8
153.20
DOCQBKMMNPJLOR-UHFFFAOYSA-N
5706487
NC(S)=C1C=CC(=O)C=C1
C7H7NOS
MFCD04973332
4-hydroxythiobenzamide, 4-hydroxybenzothioamide, 4-hydroxy thiobenzamide, 4-hydroxybenzene-1-carbothioamide, 4-hydroxybenzenecarbothioamide, benzenecarbothioamide, 4-hydroxy, 4-hydroxy-thiobenzamide, 4-aminothioxomethyl phenol, 4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one, p-hydroxythiobenzamide
4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one

Specifications

25984-63-8
C7H7NOS
4-hydroxythiobenzamide, 4-hydroxybenzothioamide, 4-hydroxy thiobenzamide, 4-hydroxybenzene-1-carbothioamide, 4-hydroxybenzenecarbothioamide, benzenecarbothioamide, 4-hydroxy, 4-hydroxy-thiobenzamide, 4-aminothioxomethyl phenol, 4-amino sulfanyl methylidene cyclohexa-2,5-dien-1-one, p-hydroxythiobenzamide
NC(S)=C1C=CC(=O)C=C1
153.20
153.2
4-Hydroxythiobenzamide
181°C to 185°C
MFCD04973332
DOCQBKMMNPJLOR-UHFFFAOYSA-N
4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
5706487
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only