missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxyoctanophenone, 99%, Thermo Scientific Chemicals

Product Code. 11362557
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2589-73-3
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.31
MDL Number MFCD00082694
InChI Key GPDYSJOGSNWMDZ-UHFFFAOYSA-N
Synonym 4-octanoylphenol, 1-4-hydroxyphenyl octan-1-one, 4'-hydroxyoctanophenone, 1-octanone, 1-4-hydroxyphenyl, octanophenone, 4'-hydroxy, unii-e2e77z6duk, e2e77z6duk, 1-4-hydroxyphenyl-1-octanone
PubChem CID 75767
IUPAC Name 1-(4-hydroxyphenyl)octan-1-one
SMILES CCCCCCCC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
Beilstein 1954746
Formula Weight 220.31
Percent Purity 99%
Chemical Name or Material 4'-Hydroxyoctanophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'