Learn More
4-(Hydroxymethyl)imidazole hydrochloride, 99%, Thermo Scientific™
48.04 € - 874.35 €
Chemical Identifiers
CAS | 32673-41-9 |
---|---|
Molecular Formula | C4H7ClN2O |
Molecular Weight (g/mol) | 134.56 |
MDL Number | MFCD00012697 |
InChI Key | WFNASTYGEKUMIY-UHFFFAOYSA-N |
Synonym | 4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride |
PubChem CID | 122926 |
IUPAC Name | 1H-imidazol-5-ylmethanol;hydrochloride |
SMILES | Cl.OCC1=CN=CN1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10327861
|
Thermo Scientific
204870010 |
1 g | Glass bottle |
48.04 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10643372
|
Thermo Scientific Acros
204870050 |
5 g | Glass bottle |
169.85 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10095813
|
Thermo Scientific Acros
204870250 |
25 g | Glass bottle | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
32673-41-9 | |
134.56 | |
WFNASTYGEKUMIY-UHFFFAOYSA-N | |
122926 | |
Cl.OCC1=CN=CN1 |
C4H7ClN2O | |
MFCD00012697 | |
4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride | |
1H-imidazol-5-ylmethanol;hydrochloride |
Specifications
32673-41-9 | |
100.0 | |
Beige to Yellow or Brown | |
99% | |
MFCD00012697 | |
4-imidazolemethanol hydrochloride, 4-hydroxymethyl imidazole hydrochloride, 1h-imidazol-4-yl methanol hydrochloride, 4-hydroxymethylimidazole hydrochloride, imidazol-4-ylmethanol hydrochloride, 1h-imidazol-4-ylmethanol hydrochloride, 1h-imidazol-5-ylmethanol hydrochloride, 4-hydroxymethyl imidazole hcl, 1h-imidazole-4-ylmethanol hydrochloride, 4 5-hydroxymethylimidazole hydrochloride | |
Cl.OCC1=CN=CN1 | |
134.56 | |
134.57 | |
Crystalline Powder and/or Chunks |
98.5 | |
110°C to 112°C | |
Authentic | |
C4H7ClN2O | |
23, I, 103 | |
WFNASTYGEKUMIY-UHFFFAOYSA-N | |
1H-imidazol-5-ylmethanol;hydrochloride | |
122926 | |
99% | |
4-(Hydroxymethyl)imidazole hydrochloride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 251-150-
- 4-(Hydroxymethyl)imidazole hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.