missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzamidine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15474157
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38148-63-9
Molecular Formula C7H9ClN2O
Molecular Weight (g/mol) 172.61
MDL Number MFCD00136405
InChI Key OADOZRQXRQAJDT-UHFFFAOYSA-N
Synonym 4-hydroxybenzamidine hydrochloride, 4-amidinophenol hydrochloride, 4-hydroxy-benzamidine hcl, p-hydroxybenzamidine hydrochloride, 4-hydroxybenzimidamide hydrochloride, 4-hydroxybenzenecarboximidamide hydrochloride, 4-hydroxybenzene-1-carboximidamide hydrochloride, benzenecarboximidamide, 4-hydroxy-, monohydrochloride, benzamidine, p-hydroxy-, monohydrochloride, 4-hydroxybenzenecarboximidamide monohydrochloride
PubChem CID 5493520
IUPAC Name 4-(diaminomethylidene)cyclohexa-2,5-dien-1-one;hydrochloride
SMILES [H+].[Cl-].NC(N)=C1C=CC(=O)C=C1

Specifications

Melting Point 221°C to 224°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 172.61
Percent Purity 95%
Chemical Name or Material 4-Hydroxybenzamidine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.