Learn More
4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals
41.00 € - 619.08 €
Chemical Identifiers
CAS | 123-08-0 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.12 |
InChI Key | RGHHSNMVTDWUBI-UHFFFAOYSA-N |
Synonym | p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde |
PubChem CID | 126 |
ChEBI | CHEBI:17597 |
IUPAC Name | 4-hydroxybenzaldehyde |
SMILES | C1=CC(=CC=C1C=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521421
|
Thermo Scientific Acros
162770500 |
50 g | Plastic bottle |
41.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184460
|
Thermo Scientific Acros
162772500 |
250 g | Plastic bottle |
103.52 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10448720
|
Thermo Scientific Acros
162770010 |
1 kg | Plastic bottle |
294.47 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954791
|
Thermo Scientific Acros
162770025 |
2.5 kg | Plastic bottle |
619.08 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-08-0 | |
122.12 | |
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
CHEBI:17597 | |
C1=CC(=CC=C1C=O)O |
C7H6O2 | |
RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
126 | |
4-hydroxybenzaldehyde |
Specifications
123-08-0 | |
Yellow to Brown | |
98.5% min. (GC) | |
HOC6H4CHO | |
15,4849 | |
Solubility in water: 13g/L (30°C). Other solubilities: 36.8g/L benzene (65°C) | |
C1=CC(=CC=C1C=O)O | |
122.12 | |
CHEBI:17597 | |
99% | |
4-Hydroxybenzaldehyde |
114.0°C to 118.0°C | |
Authentic | |
C7H6O2 | |
08,64 | |
p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde | |
RGHHSNMVTDWUBI-UHFFFAOYSA-N | |
4-hydroxybenzaldehyde | |
126 | |
122.12 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-599-1
- 4-Hydroxybenzaldehyde
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.