Learn More
4'-Hydroxyacetophenone, 99%, Thermo Scientific Chemicals
60.40 € - 233.16 €
Chemical Identifiers
CAS | 99-93-4 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00002359 |
InChI Key | TXFPEBPIARQUIG-UHFFFAOYSA-N |
Synonym | 4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone |
PubChem CID | 7469 |
ChEBI | CHEBI:28032 |
IUPAC Name | 1-(4-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314719
|
Thermo Scientific Alfa Aesar
A11974.22 |
100 g |
60.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324719
|
Thermo Scientific Alfa Aesar
A11974.36 |
500 g |
136.68 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404004
|
Thermo Scientific Alfa Aesar
A11974.0B |
1000 g |
233.16 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4?-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Hydroxyacetophenone has been used as ketone component in the preparation of 1-aryl-3-phenethylamino-1-propanone hydrochlorides, potential cytotoxic agents, via Mannich reactions.
Solubility
Soluble in 95% Ethanol. Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
99-93-4 | |
136.15 | |
TXFPEBPIARQUIG-UHFFFAOYSA-N | |
7469 | |
1-(4-hydroxyphenyl)ethanone |
C8H8O2 | |
MFCD00002359 | |
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
CHEBI:28032 | |
CC(=O)C1=CC=C(O)C=C1 |
Specifications
99-93-4 | |
286°C | |
C8H8O2 | |
774355 | |
Soluble in 95% Ethanol. Slightly soluble in water. | |
CC(=O)C1=CC=C(O)C=C1 | |
136.15 | |
CHEBI:28032 | |
99% |
106°C to 108°C | |
166°C (330°F) | |
MFCD00002359 | |
4'-hydroxyacetophenone, 4-hydroxyacetophenone, 1-4-hydroxyphenyl ethanone, 4-acetylphenol, p-hydroxyacetophenone, piceol, ethanone, 1-4-hydroxyphenyl, p-acetylphenol, para-hydroxyacetophenone, p-oxyacetophenone | |
TXFPEBPIARQUIG-UHFFFAOYSA-N | |
1-(4-hydroxyphenyl)ethanone | |
7469 | |
136.15 | |
4'-Hydroxyacetophenone |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 202-802-8
missing translation for 'rtecsNumber' : AM8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 4'-Hydroxyacetophenone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : p-Hydroxyacetophenone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.