missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethoxy)benzaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 15468135
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53104-95-3
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD04973780
InChI Key GPJSLRQIOKFRFL-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethoxy benzaldehyde, benzaldehyde, 4-hydroxy-3-trifluoromethoxy, 4-hydroxy-3-trifluoromethoxy benzaldehyde, 98+%, 4-oxidanyl-3-trifluoromethyloxy benzaldehyde
PubChem CID 7018050
IUPAC Name 4-hydroxy-3-(trifluoromethoxy)benzaldehyde
SMILES OC1=C(OC(F)(F)F)C=C(C=O)C=C1

Specifications

Melting Point 76°C to 80°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 206.12
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethoxy)benzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.