Learn More
4-Hydroxy-3,5-dimethoxycinnamic acid, 98%, Thermo Scientific Chemicals
30.40 € - 358.00 €
Chemical Identifiers
CAS | 530-59-6 |
---|---|
Molecular Formula | C11H12O5 |
Molecular Weight (g/mol) | 224.212 |
MDL Number | MFCD00004401 |
InChI Key | PCMORTLOPMLEFB-ONEGZZNKSA-N |
Synonym | sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec |
PubChem CID | 637775 |
ChEBI | CHEBI:15714 |
IUPAC Name | (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid |
SMILES | COC1=CC(=CC(=C1O)OC)C=CC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379519
|
Thermo Scientific Alfa Aesar
A15676.03 |
1 g |
30.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457617
|
Thermo Scientific Alfa Aesar
A15676.06 |
5 g |
89.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467617
|
Thermo Scientific Alfa Aesar
A15676.14 |
25 g |
358.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.
Solubility
Soluble in methanol, ethanol and dimethyl sulfoxide.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Chemical Identifiers
530-59-6 | |
224.212 | |
PCMORTLOPMLEFB-ONEGZZNKSA-N | |
637775 | |
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid |
C11H12O5 | |
MFCD00004401 | |
sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec | |
CHEBI:15714 | |
COC1=CC(=CC(=C1O)OC)C=CC(=O)O |
Specifications
530-59-6 | |
C11H12O5 | |
2130006 | |
Soluble in methanol,ethanol and dimethyl sulfoxide. | |
COC1=CC(=CC(=C1O)OC)C=CC(=O)O | |
224.212 | |
CHEBI:15714 | |
98% |
∼195°C (decomposition) | |
MFCD00004401 | |
sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec | |
PCMORTLOPMLEFB-ONEGZZNKSA-N | |
(E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid | |
637775 | |
224.21 | |
4-Hydroxy-3,5-dimethoxycinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 208-487-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only