missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-1-indanone, 97%, Thermo Scientific™
99.20 € - 321.00 €
Chemical Identifiers
CAS | 40731-98-4 |
---|---|
Molecular Formula | C9H8O2 |
Molecular Weight (g/mol) | 148.16 |
MDL Number | MFCD00143330 |
InChI Key | CKSCMRNFDBWFND-UHFFFAOYSA-N |
Synonym | 4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659 |
PubChem CID | 590547 |
IUPAC Name | 4-hydroxy-2,3-dihydroinden-1-one |
SMILES | OC1=CC=CC2=C1CCC2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10602255
|
Thermo Scientific Acros
399030010 |
1 g |
99.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10366394
|
Thermo Scientific Acros
399030050 |
5 g |
321.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
40731-98-4 | |
148.16 | |
CKSCMRNFDBWFND-UHFFFAOYSA-N | |
590547 | |
OC1=CC=CC2=C1CCC2=O |
C9H8O2 | |
MFCD00143330 | |
4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659 | |
4-hydroxy-2,3-dihydroinden-1-one |
Specifications
40731-98-4 | |
96% min. (HPLC) | |
C9H8O2 | |
4-hydroxy-1-indanone, 4-hydroxyindan-1-one, 4-hydroxyindanone, 4-hydroxy-indan-1-one, 4-hydroxy-2,3-dihydro-1h-inden-1-one, 2,3-dihydro-4-hydroxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-4-hydroxy, 2,3-dihydro-4-hydroxy-1h-inden-1-one, 4-hydroxy-indanone, pubchem9659 | |
OC1=CC=CC2=C1CCC2=O | |
148.16 | |
148.16 | |
4-Hydroxy-1-indanone |
242°C to 244°C | |
Glass bottle | |
MFCD00143330 | |
CKSCMRNFDBWFND-UHFFFAOYSA-N | |
4-hydroxy-2,3-dihydroinden-1-one | |
590547 | |
97% |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Hydroxy-1-indanone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title