missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenyl beta-D-allopyranoside, 98%, Thermo Scientific Chemicals

33.80 € - 57.69 €

Chemical Identifiers

CAS 80154-34-3
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.264
MDL Number MFCD00210992
InChI Key OLZAGZCCJJBKNZ-SYLRKERUSA-N
Synonym helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
PubChem CID 12896796
IUPAC Name 4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15417339
View Documents
Thermo Scientific Alfa Aesar
J66102.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15427339
View Documents
Thermo Scientific Alfa Aesar
J66102.06
5 g
57.69 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in the treatment of ischemic cardiovascular and cerebrovascular diseases. Also used in Chinese medicine in the treatment of headache and insomnia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water. Slightly soluble in water, ethanol or carbinol. Insoluble in aether or chloroform.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

80154-34-3
284.264
OLZAGZCCJJBKNZ-SYLRKERUSA-N
12896796
C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
C13H16O7
MFCD00210992
helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde

Specifications

80154-34-3
White
MFCD00210992
Soluble in hot water. Slightly soluble in water,ethanol or carbinol. Insoluble in aether or chloroform.
−105° (c=0.8 in Water)
4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
12896796
98%
4-Formylphenyl beta-D-allopyranoside
190°C
C13H16O7
helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
OLZAGZCCJJBKNZ-SYLRKERUSA-N
C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O
284.264
284.26
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.